Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAM9
Entrez Gene ID
397344
Gene Name
ADAM metallopeptidase domain 9
Gene Aliases
ADAM-9
Chromosome Location
Chr.15: 47523768 - 47601722 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001925664.5 XP_001925699.2 - 1844 96
XM_021075120.1 XP_020930779.1 - 1850 96
GenBank mRNA AF069646.1 - 2 - 3 271 96
AK352011.1 - 14 - 15 1703 96
AK394001.1 - 14 - 15 1790 96

View Full Product Details