Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RXRG
Entrez Gene ID
445465
Gene Name
retinoid X receptor gamma
Gene Aliases
NR2B3, RXRgamma
Chromosome Location
Chr.4: 85158313 - 85357116 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130213.1 NP_001123685.1 2 - 3 464 75
XM_013988076.2 XP_013843530.1 - 1122 75
XM_021088737.1 XP_020944396.1 - 517 75
GenBank mRNA AY429474.1 - 1 - 2 199 75
DQ866834.1 - 2 - 3 464 75

View Full Product Details