Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC27A2
Entrez Gene ID
494011
Gene Name
solute carrier family 27 member 2
Gene Aliases
ACSVL1, FACVL1, HsT17226, hFACVL1
Chromosome Location
Chr.1: 121530449 - 121578994 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278777.1 NP_001265706.1 7 - 8 1537 117
GenBank mRNA AK233289.1 - 7 - 8 1545 117
AY822678.1 - 3 - 4 320 117
AY822679.1 - 1 - 2 137 117
JX092264.1 - 7 - 8 1537 117

View Full Product Details