Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AQP6
Entrez Gene ID
100145893
Gene Name
aquaporin 6
Gene Aliases
-
Chromosome Location
Chr.5: 15888080 - 15892113 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128467.1 NP_001121939.1 2 - 3 547 71
XM_021090894.1 XP_020946553.1 - 726 71
GenBank mRNA EU620575.1 - 2 - 3 547 71

View Full Product Details