Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CYP27A1
Entrez Gene ID
100126282
Gene Name
cytochrome P450, family 27, subfamily A, polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.15: 120809886 - 120852196 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243304.1 NP_001230233.1 9 - 9 2452 94
GenBank mRNA AK232936.1 - 9 - 9 2452 94
AK392950.1 - 9 - 9 1571 94

View Full Product Details