Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
CXCL14
Entrez Gene ID
494467
Gene Name
C-X-C motif chemokine ligand 14
Gene Aliases
BRAK
Chromosome Location
Chr.2: 137803988 - 137813118 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY308800.1 - 1 - 1 48 79

View Full Product Details