Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
KCNS3
Entrez Gene ID
733666
Gene Name
potassium voltage-gated channel modifier subfamily S member 3
Gene Aliases
-
Chromosome Location
Chr.3: 119951110 - 119989541 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044596.1 NP_001038061.1 4 - 4 1854 85
XM_021085911.1 XP_020941570.1 - 2077 85
XM_021085912.1 XP_020941571.1 - 1941 85
XM_021085913.1 XP_020941572.1 - 2130 85
XM_021085915.1 XP_020941574.1 - 1980 85
XM_021085916.1 XP_020941575.1 - 1971 85
XM_021085917.1 XP_020941576.1 - 1782 85
XM_021085918.1 XP_020941577.1 - 2700 85
GenBank mRNA AK344049.1 - 3 - 3 1463 85
DQ005443.1 - 4 - 4 1854 85

View Full Product Details