Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FASLG
Entrez Gene ID
396726
Gene Name
Fas ligand (TNF superfamily, member 6)
Gene Aliases
CD95-L, FASL, TNFSF6
Chromosome Location
Chr.9: 115068314 - 115075147 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213806.1 NP_998971.1 4 - 4 651 95
GenBank mRNA AB027297.1 - 4 - 4 651 95
AF121892.1 - 4 - 4 650 95

View Full Product Details