Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CYP3A29
Entrez Gene ID
403324
Gene Name
cytochrome P450 3A29
Gene Aliases
-
Chromosome Location
Chr.3: 6738502 - 6778121 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214423.1 NP_999588.1 11 - 11 1254 63
GenBank mRNA AF424780.1 - 11 - 11 1188 63
AK232311.1 - 11 - 11 1267 63
AY610066.1 - - 1250 63
EU918131.1 - 11 - 11 1223 63
Z93099.1 - - 1254 63

View Full Product Details