Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
PR39
Entrez Gene ID
407610
Gene Name
peptide antibiotic PR39
Gene Aliases
PR-39
Chromosome Location
Chr.13: 30937930 - 30957127 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214450.1 NP_999615.1 4 - 4 452 146

View Full Product Details