Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFIP11
Entrez Gene ID
768099
Gene Name
tuftelin interacting protein 11
Gene Aliases
STIP
Chromosome Location
Chr.14: 44097026 - 44113760 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077214.1 NP_001070682.1 10 - 11 1897 103
XM_021072205.1 XP_020927864.1 - 1962 103
XM_021072206.1 XP_020927865.1 - 2030 103
GenBank mRNA DQ923315.1 - 10 - 11 1897 103

View Full Product Details