Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC42
Entrez Gene ID
780428
Gene Name
cell division cycle 42
Gene Aliases
-
Chromosome Location
Chr.6: 80035034 - 80089243 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078680.1 NP_001072148.1 4 - 5 486 113
XM_005656040.3 XP_005656097.1 - 653 113
GenBank mRNA DQ915496.1 - 4 - 5 486 113

View Full Product Details