Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CBLN3
Entrez Gene ID
100127149
Gene Name
cerebellin 3 precursor
Gene Aliases
-
Chromosome Location
Chr.7: 74865931 - 74868443 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112680.1 NP_001106150.1 2 - 3 525 67
GenBank mRNA EF577505.1 - 2 - 3 525 67

View Full Product Details