Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLA-DRB1
Entrez Gene ID
100153386
Gene Name
MHC class II histocompatibility antigen SLA-DRB1
Gene Aliases
DRB1, LA-DRB-c, LA-DRB-d, SLA-DRB, SLADRB
Chromosome Location
Chr.7: 24901386 - 24914016 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113695.1 NP_001107167.1 5 - 6 860 117
GenBank mRNA AB016746.1 - 4 - 5 701 117
AF464046.2 - 5 - 6 781 117
AK348716.1 - 5 - 6 845 117
AK394000.1 - 5 - 6 853 117
AK396034.1 - 5 - 6 852 117
AK396070.1 - 5 - 6 856 117
AY135576.1 - 4 - 5 498 117
AY135579.1 - 5 - 6 860 117
AY135580.1 - 5 - 6 748 117
DQ883227.1 - 5 - 6 809 117
EU432072.1 - 5 - 6 790 117
EU432076.1 - 5 - 6 790 117
EU432079.1 - 5 - 6 790 117
KU754598.1 - 5 - 6 809 117
LC020542.1 - 5 - 6 117
LC020543.1 - 5 - 6 117

Gene Symbol
LOC106504372
Entrez Gene ID
106504372
Gene Name
DLA class II histocompatibility antigen, DR-1 beta chain-like
Gene Aliases
DRB1, SLA-DRB
Chromosome Location
Chr.7: 25000257 - 25000539 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK396361.1 - 5 - 6 878 117

View Full Product Details