Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TUBB1
Entrez Gene ID
100134983
Gene Name
tubulin beta 1 class VI
Gene Aliases
-
Chromosome Location
Chr.17: 59161420 - 59168385 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113696.1 NP_001107168.1 3 - 4 466 60

View Full Product Details