Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFNB2
Entrez Gene ID
100135673
Gene Name
ephrin B2
Gene Aliases
-
Chromosome Location
Chr.11: 74188337 - 74232768 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114286.1 NP_001107758.1 3 - 4 493 116
GenBank mRNA EF682141.1 - 3 - 4 493 116
KF500033.1 - 3 - 4 581 116

View Full Product Details