Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LSM2
Entrez Gene ID
100144534
Gene Name
LSM2 homolog, U6 small nuclear RNA and mRNA degradation associated
Gene Aliases
-
Chromosome Location
Chr.7: 23900385 - 23908523 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123136.1 NP_001116608.1 2 - 3 296 154
GenBank mRNA AK233908.1 - 2 - 3 296 154
AK346706.1 - 2 - 3 299 154
AK346872.1 - 2 - 3 276 154
AK347071.1 - 2 - 3 304 154

View Full Product Details