Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UPK2
Entrez Gene ID
397075
Gene Name
uroplakin 2
Gene Aliases
UP2, UPII
Chromosome Location
Chr.9: 46200465 - 46202754 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214012.1 NP_999177.1 4 - 5 446 71
XM_021102209.1 XP_020957868.1 - 496 71
GenBank mRNA AF164141.2 - 4 - 5 446 71

View Full Product Details