Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GP91-PHOX
Entrez Gene ID
397108
Gene Name
NADPH oxidase heavy chain subunit
Gene Aliases
CYBB, NOX2
Chromosome Location
Chr.X: 33654656 - 33678346 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214043.2 NP_999208.1 3 - 4 412 125
GenBank mRNA AK230821.1 - 6 - 7 125
AK231643.1 - 6 - 7 744 125
AK232201.1 - 6 - 7 125
AK345180.1 - 6 - 7 744 125
AK398894.1 - 6 - 7 723 125
AK398895.1 - 6 - 7 723 125
U02476.1 - 3 - 4 412 125

View Full Product Details