Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BPIFA1
Entrez Gene ID
449524
Gene Name
BPI fold containing family A member 1
Gene Aliases
PLUNC, SPLUNC1
Chromosome Location
Chr.17: 36754498 - 36761622 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005727.1 NP_001005727.1 3 - 4 416 76
XM_005672834.1 XP_005672891.1 - 595 76
XM_005672835.2 XP_005672892.1 - 491 76
GenBank mRNA AK238135.1 - 4 - 5 473 76
AK238161.1 - 4 - 5 494 76
AK238256.1 - 4 - 5 473 76
AK238296.1 - 4 - 5 473 76
AK350801.1 - 3 - 4 460 76
AK350937.1 - 4 - 5 474 76
AK397439.1 - 4 - 5 473 76
AK397446.1 - 4 - 5 473 76
AK397447.1 - 4 - 5 474 76
AK397448.1 - 4 - 5 473 76
AK397451.1 - 4 - 5 473 76
AK397460.1 - 4 - 5 473 76
AK397478.1 - 4 - 5 476 76
AK397484.1 - 4 - 5 473 76
AK397490.1 - 4 - 5 595 76
AK397496.1 - 4 - 5 499 76
AK397506.1 - 4 - 5 473 76
AK397531.1 - 4 - 5 498 76
AK397535.1 - 4 - 5 473 76
AK397538.1 - 4 - 5 473 76
AK397555.1 - 4 - 5 479 76
AK397556.1 - 4 - 5 473 76
AK397606.1 - 4 - 5 473 76
AK397619.1 - 4 - 5 477 76
AK397620.1 - 4 - 5 478 76
AK397909.1 - 4 - 5 475 76
AK397956.1 - 4 - 5 479 76
AK397957.1 - 4 - 5 475 76
AK397999.1 - 4 - 5 479 76
AK398049.1 - 4 - 5 480 76
AK398104.1 - 4 - 5 475 76
AK399196.1 - 4 - 5 473 76
AK399197.1 - 4 - 5 504 76
AK399199.1 - 4 - 5 476 76
AK399201.1 - 4 - 5 473 76
AK399203.1 - 4 - 5 473 76
AK399205.1 - 4 - 5 473 76
AK399208.1 - 4 - 5 477 76
AK399209.1 - 4 - 5 478 76
AK399210.1 - 4 - 5 477 76
AK399218.1 - 4 - 5 477 76
AK399221.1 - 4 - 5 473 76
AK399225.1 - 4 - 5 483 76
AK399227.1 - 4 - 5 489 76
AK399228.1 - 4 - 5 477 76
AK399233.1 - 4 - 5 473 76
AK399234.1 - 4 - 5 478 76
AK399236.1 - 4 - 5 473 76
AK399238.1 - 4 - 5 473 76
AK399241.1 - 4 - 5 473 76
AK399242.1 - 4 - 5 473 76
AK399245.1 - 4 - 5 473 76
AK399248.1 - 4 - 5 473 76
AK399252.1 - 4 - 5 479 76
AK399256.1 - 4 - 5 475 76
AK399257.1 - 4 - 5 477 76
AK399259.1 - 4 - 5 473 76
AK399260.1 - 4 - 5 481 76
AK399261.1 - 4 - 5 473 76
AK399334.1 - 4 - 5 475 76
AK399340.1 - 4 - 5 476 76
AK399350.1 - 4 - 5 475 76
AK399351.1 - 4 - 5 483 76
AY733063.1 - 3 - 4 416 76

View Full Product Details