Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NR1I3
Entrez Gene ID
654317
Gene Name
nuclear receptor subfamily 1 group I member 3
Gene Aliases
CAR
Chromosome Location
Chr.4: 89226314 - 89231781 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037996.1 NP_001033085.1 2 - 3 319 61
XM_013996822.2 XP_013852276.1 - 597 61
GenBank mRNA AB214979.1 - 2 - 3 319 61
AK232831.1 - 2 - 3 319 61
AK346033.1 - 2 - 3 319 61
DQ249306.1 - 1 - 2 109 61
KF586862.1 - 1 - 2 103 61
KF586863.1 - 1 - 2 103 61
KF586864.1 - 1 - 2 103 61
KF586865.1 - 1 - 2 103 61
KF586866.1 - 1 - 2 103 61

View Full Product Details