Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPARD
Entrez Gene ID
397297
Gene Name
peroxisome proliferator activated receptor delta
Gene Aliases
PPARbeta
Chromosome Location
Chr.7: 31222533 - 31295221 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130241.3 NP_001123713.1 3 - 4 514 82
NM_214152.3 NP_999317.2 4 - 5 598 82
XM_005665910.3 XP_005665967.1 - 679 82
XM_013977808.2 XP_013833262.1 - 1303 82
GenBank mRNA AF228697.1 - 2 - 3 335 82
AY188501.1 - 4 - 5 564 82
DQ437886.1 - 3 - 4 514 82
GU565977.1 - 2 - 3 384 82
KP757025.1 - 2 - 3 283 82
KP757026.1 - 2 - 3 283 82

View Full Product Details