Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIGLEC1
Entrez Gene ID
397623
Gene Name
sialic acid binding Ig like lectin 1
Gene Aliases
SA, SIGLEC-1, SN, p210, pSn
Chromosome Location
Chr.17: 31994505 - 32021239 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214346.1 NP_999511.1 10 - 11 2680 89
XM_021077303.1 XP_020932962.1 - 2944 89
XM_021077304.1 XP_020932963.1 - 2946 89
GenBank mRNA AF509585.1 - 10 - 11 2680 89
DQ176853.1 - 10 - 11 2680 89
EU003993.1 - 10 - 11 2680 89
JX070181.1 - 10 - 11 2680 89
JX070182.1 - 10 - 11 2680 89
JX070183.1 - 10 - 11 2680 89

View Full Product Details