Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- IFNA1
- Entrez Gene ID
-
397686
- Gene Name
- interferon, alpha 1
- Gene Aliases
-
IFN-ALPHA-1, IFN1@
- Chromosome Location
-
Chr.1: 201515835 - 201516477 on Build Sscrofa11.1
- Gene Symbol
- IFN-ALPHA-1
- Entrez Gene ID
-
100157352
- Gene Name
- interferon-alpha-1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 201290138 - 201290683 on Build Sscrofa11.1
- Gene Symbol
- IFN-ALPHA-10
- Entrez Gene ID
-
100156561
- Gene Name
- interferon-alpha-10
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 201353877 - 201354422 on Build Sscrofa11.1
- Gene Symbol
- IFN-ALPHA-13
- Entrez Gene ID
-
100152100
- Gene Name
- interferon-alpha-13
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 201770996 - 201771565 on Build Sscrofa11.1
- Gene Symbol
- IFN-ALPHA-14
- Entrez Gene ID
-
100310804
- Gene Name
- interferon-alpha-14
- Gene Aliases
-
IFN-alpha
- Chromosome Location
-
Chr.1: 201749695 - 201750264 on Build Sscrofa11.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JQ839262.1 |
- |
1 - 1 |
466 |
101 |
- Gene Symbol
- IFN-ALPHA-15
- Entrez Gene ID
-
100152504
- Gene Name
- interferon-alpha-15
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 201694271 - 201694837 on Build Sscrofa11.1
- Gene Symbol
- IFN-ALPHA-17
- Entrez Gene ID
-
100154938
- Gene Name
- interferon-alpha-17
- Gene Aliases
-
IFNa8
- Chromosome Location
-
Chr.1: 201321905 - 201322450 on Build Sscrofa11.1
- Gene Symbol
- IFN-ALPHA-4
- Entrez Gene ID
-
100310805
- Gene Name
- interferon-alpha-4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 201616020 - 201616589 on Build Sscrofa11.1
- Gene Symbol
- IFN-ALPHA-5
- Entrez Gene ID
-
100303758
- Gene Name
- interferon, alpha 5
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 201535293 - 201535862 on Build Sscrofa11.1
- Gene Symbol
- IFN-ALPHA-8
- Entrez Gene ID
-
100153724
- Gene Name
- interferon-alpha-8
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 201391451 - 201392020 on Build Sscrofa11.1
- Gene Symbol
- IFN-ALPHA-9
- Entrez Gene ID
-
100157766
- Gene Name
- interferon-alpha-9
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 201407647 - 201408216 on Build Sscrofa11.1
- Gene Symbol
- LOC100153325
- Entrez Gene ID
-
100153325
- Gene Name
- interferon alpha-2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 201272569 - 201273114 on Build Sscrofa11.1
- Gene Symbol
- LOC100153661
- Entrez Gene ID
-
100153661
- Gene Name
- interferon alpha-14
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 201736733 - 201737300 on Build Sscrofa11.1
- Gene Symbol
- LOC100156936
- Entrez Gene ID
-
100156936
- Gene Name
- interferon alpha-11
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 201456457 - 201457002 on Build Sscrofa11.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
GQ415061.1 |
- |
1 - 1 |
416 |
101 |
- Gene Symbol
- LOC100737904
- Entrez Gene ID
-
100737904
- Gene Name
- interferon alpha-1-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 201261017 - 201262150 on Build Sscrofa11.1
View Full Product Details