Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PGA5
Entrez Gene ID
396892
Gene Name
pepsinogen 5, group I (pepsinogen A)
Gene Aliases
PGA
Chromosome Location
Chr.2: 10244300 - 10252644 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213873.2 NP_999038.2 3 - 4 352 65
GenBank mRNA EF108301.1 - 3 - 4 352 65
J04601.1 - 3 - 4 369 65
M20920.1 - 3 - 4 405 65

View Full Product Details