Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PGA5
Entrez Gene ID
396892
Gene Name
pepsinogen 5, group I (pepsinogen A)
Gene Aliases
PGA
Chromosome Location
Chr.2: 10244300 - 10252644 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213873.2 NP_999038.2 5 - 6 668 62
GenBank mRNA EF108301.1 - 5 - 6 668 62
M20920.1 - 5 - 6 721 62

View Full Product Details