Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SGCA
Entrez Gene ID
100240723
Gene Name
sarcoglycan alpha
Gene Aliases
-
Chromosome Location
Chr.12: 26356691 - 26368561 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144122.1 NP_001137594.1 7 - 7 853 73
XM_021066044.1 XP_020921703.1 - 974 73
GenBank mRNA FJ481167.1 - 7 - 7 853 73

View Full Product Details