The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167654.1 | NP_001161126.1 | 1 - 1 | 747 | 87 |
XM_005667778.3 | XP_005667835.1 | - | 1249 | 87 | |
XM_005667779.3 | XP_005667836.1 | - | 1203 | 87 | |
GenBank mRNA | AB529870.1 | - | 1 - 1 | 747 | 87 |
AK231548.1 | - | 3 - 3 | 1254 | 87 |