Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPACA5
Entrez Gene ID
100415934
Gene Name
sperm acrosome associated 5
Gene Aliases
-
Chromosome Location
Chr.X: 42476719 - 42478783 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177930.1 NP_001171401.1 2 - 3 300 84
GenBank mRNA GU474993.1 - 2 - 3 300 84

View Full Product Details