Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DOK2
Entrez Gene ID
100152025
Gene Name
docking protein 2
Gene Aliases
-
Chromosome Location
Chr.14: 6176487 - 6180895 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185102.1 NP_001172031.1 4 - 5 687 70
GenBank mRNA AK231458.1 - 4 - 5 689 70

View Full Product Details