Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NANS
Entrez Gene ID
100155883
Gene Name
N-acetylneuraminate synthase
Gene Aliases
-
Chromosome Location
Chr.1: 239881984 - 239912727 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185139.1 NP_001172068.1 5 - 6 873 56
GenBank mRNA AB529862.1 - 5 - 6 873 56
AK240367.1 - 5 - 6 984 56
AK347608.1 - 5 - 6 947 56
AY609633.1 - 5 - 6 922 56

View Full Product Details