Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERH
Entrez Gene ID
100152505
Gene Name
ERH mRNA splicing and mitosis factor
Gene Aliases
-
Chromosome Location
Chr.7: 93068254 - 93083950 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185163.1 NP_001172092.1 2 - 3 152 104
GenBank mRNA AK392587.1 - 2 - 3 171 104
AK399827.1 - 2 - 3 177 104
AY610375.1 - 2 - 3 122 104

View Full Product Details