Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM204A
Entrez Gene ID
100152540
Gene Name
family with sequence similarity 204 member A
Gene Aliases
C10orf84, C14H10orf84
Chromosome Location
Chr.14: 128366152 - 128402587 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185171.1 NP_001172100.1 2 - 3 266 95

View Full Product Details