Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR45
Entrez Gene ID
100462757
Gene Name
WD repeat domain 45
Gene Aliases
-
Chromosome Location
Chr.X: 43150691 - 43155981 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190293.1 NP_001177222.1 10 - 11 1106 88
XM_013986108.2 XP_013841562.1 - 1163 88
XM_013986109.2 XP_013841563.1 - 1151 88
XM_013986110.2 XP_013841564.1 - 1283 88
XM_013986111.2 XP_013841565.1 - 1229 88
GenBank mRNA AK235058.1 - 10 - 11 1102 88
AK398278.1 - 10 - 11 1109 88
HM046518.1 - 10 - 11 1106 88

View Full Product Details