Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAT9
Entrez Gene ID
100359356
Gene Name
N-acetyltransferase 9 (putative)
Gene Aliases
-
Chromosome Location
Chr.12: 6459673 - 6464236 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195360.1 NP_001182289.1 5 - 6 504 89
XM_005668615.3 XP_005668672.1 - 1240 89
XM_013980760.2 XP_013836214.1 - 485 89
XM_021066064.1 XP_020921723.1 - 474 89
GenBank mRNA GU373685.1 - 5 - 6 504 89

View Full Product Details