Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPM2
Entrez Gene ID
100154488
Gene Name
nucleophosmin/nucleoplasmin 2
Gene Aliases
-
Chromosome Location
Chr.14: 6288422 - 6303750 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195362.1 NP_001182291.1 6 - 7 660 71
XM_005657214.3 XP_005657271.2 - 653 71
XM_005657216.3 XP_005657273.2 - 1531 71
GenBank mRNA GU373718.1 - 6 - 7 660 71

View Full Product Details