Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAGE2B
Entrez Gene ID
100359360
Gene Name
PAGE family member 2B
Gene Aliases
-
Chromosome Location
Chr.X: 47916076 - 47919904 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195364.1 NP_001182293.1 4 - 5 449 86
GenBank mRNA GU373676.1 - 4 - 5 449 86

View Full Product Details