Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLIC5
Entrez Gene ID
100154589
Gene Name
chloride intracellular channel 5
Gene Aliases
-
Chromosome Location
Chr.7: 40683044 - 40808017 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198923.2 NP_001185852.2 2 - 3 533 80
GenBank mRNA AK393630.1 - 2 - 3 785 80
AK395655.1 - 2 - 3 505 80
AK400732.1 - 2 - 3 532 80
EU715290.1 - 2 - 3 462 80

View Full Product Details