Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A24
Entrez Gene ID
100518780
Gene Name
solute carrier family 25 member 24
Gene Aliases
SCAMC-1
Chromosome Location
Chr.4: 111580186 - 111632390 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199363.1 NP_001186292.1 3 - 4 392 92
GenBank mRNA EU617320.1 - 3 - 4 392 92

View Full Product Details