Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VBP1
Entrez Gene ID
100514622
Gene Name
VHL binding protein 1
Gene Aliases
-
Chromosome Location
Chr.X: 125485357 - 125506781 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244980.1 NP_001231909.1 2 - 3 230 106
XM_005674019.3 XP_005674076.1 - 333 106
GenBank mRNA AK230843.1 - 2 - 3 230 106
AK392033.1 - 2 - 3 243 106
AK399546.1 - 2 - 3 253 106

View Full Product Details