Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB6A
Entrez Gene ID
100625022
Gene Name
RAB6A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.9: 8137901 - 8199212 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243273.1 NP_001230202.1 4 - 5 706 95
XM_021102115.1 XP_020957774.1 - 268 95
GenBank mRNA AK232771.1 - 4 - 5 706 95
AK346415.1 - 5 - 6 622 95
AY609534.1 - 4 - 5 441 95

View Full Product Details