Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TMEM86B
- Entrez Gene ID
-
100514582
- Gene Name
- transmembrane protein 86B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 59424127 - 59428641 on Build Sscrofa11.1
- Gene Symbol
- SART3
- Entrez Gene ID
-
100152586
- Gene Name
- spliceosome associated factor 3, U4/U6 recycling protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.14: 42223752 - 42259992 on Build Sscrofa11.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK346497.1 |
- |
- |
66 |
80 |
View Full Product Details