Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPIL6
Entrez Gene ID
100157379
Gene Name
peptidylprolyl isomerase like 6
Gene Aliases
-
Chromosome Location
Chr.1: 75465290 - 75508250 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243664.1 NP_001230593.1 2 - 3 252 102
XM_021101116.1 XP_020956775.1 - 244 102
GenBank mRNA AK345353.1 - 2 - 3 252 102
DY419200.1 - 2 - 3 185 102

View Full Product Details