Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB9
Entrez Gene ID
100158057
Gene Name
ankyrin repeat and SOCS box containing 9
Gene Aliases
-
Chromosome Location
Chr.X: 11800546 - 11828251 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243703.1 NP_001230632.1 3 - 4 528 61
XM_005673415.3 XP_005673472.1 - 680 61
XM_005673416.3 XP_005673473.1 - 680 61
XM_005673417.3 XP_005673474.1 - 572 61
XM_005673418.3 XP_005673475.1 - 572 61
XM_013985902.2 XP_013841356.1 - 703 61
GenBank mRNA AK351224.1 - 3 - 4 528 61

View Full Product Details