Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDIP1
Entrez Gene ID
100512098
Gene Name
cell death inducing p53 target 1
Gene Aliases
C16orf5, C3H16orf5
Chromosome Location
Chr.3: 37815259 - 37839266 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244099.1 NP_001231028.1 1 - 2 60 73
GenBank mRNA BE030992.1 - 1 - 2 60 73

View Full Product Details