Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDIP1
Entrez Gene ID
100512098
Gene Name
cell death inducing p53 target 1
Gene Aliases
C16orf5, C3H16orf5
Chromosome Location
Chr.3: 37815259 - 37839266 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244099.1 NP_001231028.1 4 - 5 388 114
XM_005662155.3 XP_005662212.1 - 508 114
XM_021085649.1 XP_020941308.1 - 734 114
XM_021085650.1 XP_020941309.1 - 736 114
XM_021085651.1 XP_020941310.1 - 636 114
XM_021085652.1 XP_020941311.1 - 605 114
XM_021085653.1 XP_020941312.1 - 606 114
GenBank mRNA BE030992.1 - 4 - 5 388 114

View Full Product Details