Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORD
Entrez Gene ID
100158181
Gene Name
sorbitol dehydrogenase
Gene Aliases
SDH, XDH
Chromosome Location
Chr.1: 126659198 - 126701084 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244162.1 NP_001231091.1 6 - 7 769 79
XM_021062686.1 XP_020918345.1 - 631 79
GenBank mRNA AK236025.1 - 6 - 7 729 79
AK236181.1 - 5 - 6 684 79
AK351174.1 - 5 - 6 651 79
AK389436.1 - 6 - 7 769 79
AK392246.1 - 6 - 7 757 79
AY610034.1 - 6 - 7 684 79

View Full Product Details