Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WIPF2
Entrez Gene ID
100525252
Gene Name
WAS/WASL interacting protein family member 2
Gene Aliases
-
Chromosome Location
Chr.12: 22119134 - 22164641 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244312.1 NP_001231241.1 1 - 2 134 121
XM_005653906.3 XP_005653963.1 - 433 121
XM_005653907.3 XP_005653964.1 - 349 121
XM_021066132.1 XP_020921791.1 - 304 121
XM_021066134.1 XP_020921793.1 - 276 121
GenBank mRNA CK458671.1 - 2 - 3 240 121

View Full Product Details