Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WIPF2
Entrez Gene ID
100525252
Gene Name
WAS/WASL interacting protein family member 2
Gene Aliases
-
Chromosome Location
Chr.12: 22119134 - 22164641 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244312.1 NP_001231241.1 6 - 7 1354 101
XM_005653906.3 XP_005653963.1 - 1653 101
XM_005653907.3 XP_005653964.1 - 1569 101
XM_021066132.1 XP_020921791.1 - 1524 101
XM_021066134.1 XP_020921793.1 - 1496 101
GenBank mRNA CX057944.1 - 3 - 4 567 101
DN107192.1 - 2 - 3 180 101

View Full Product Details