Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLP2
Entrez Gene ID
100512511
Gene Name
proteolipid protein 2
Gene Aliases
-
Chromosome Location
Chr.X: 43215372 - 43218318 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244435.1 NP_001231364.1 4 - 5 597 73
GenBank mRNA AK234097.1 - 4 - 5 597 73
AY609710.1 - 4 - 5 535 73
BI399865.1 - 1 - 2 423 73

View Full Product Details